Learn More
Teknova YPD Broth. 500mL, Sterile.
SDP
Catalog No. 502388204
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-238-8204 Supplier Teknova Supplier No. Y5005
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.