Learn More
Research Products International Corp YPD Broth, 25 Gram Capsules, 1 Kilogram
SDP
Catalog No. 50489176
Encompass_Preferred
Add 2 capsules to one liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes.
Ingredients:
Peptone 20 g/l
Dextrose 20 g/l
Yeast Extract 10 g/l
Catalog No. 50-489-176 Supplier Research Products International Corp Supplier No. Y200921000.0
Add 2 capsules to one liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes.
Ingredients:
Peptone 20 g/l
Dextrose 20 g/l
Yeast Extract 10 g/l