Learn More
Research Products International Corp YPD Broth, 1 Gram Capsules, 500 Grams
SDP
Catalog No. 50489175
Encompass_Preferred
Add 50 capsules to 1 liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes.
Ingredients:
Peptone 20 g/l
Dextrose 20 g/l
Yeast Extract 10 g/l
Catalog No. 50-489-175 Supplier Research Products International Corp Supplier No. Y20091500.0
Add 50 capsules to 1 liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes.
Ingredients:
Peptone 20 g/l
Dextrose 20 g/l
Yeast Extract 10 g/l