Learn More
Teknova YPD Broth with 1M Sorbitol, 1000mL, Sterile.
SDP
Catalog No. 50843582
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-843-582 Supplier Teknova Supplier No. Y5300
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.