Learn More
Teknova YPAD Broth with 0.006% Adenine Hemisulfate. 1000mL, Sterile.
SDP
Catalog No. 50843566
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-843-566 Supplier Teknova Supplier No. Y5030
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.