Learn More
Teknova YPD Broth with Animal-Free Soytone. 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387530
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-238-7530 Supplier Teknova Supplier No. Y12002PK
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.