Learn More
HIMEDIA YPD Broth (YEPD Broth), M1363, Microbiology Culture Media, Dehydrated Culture Media, Broth, Saccharomyces cerevisiae Growth, Molecular Biology, Powder, 500 g
Catalog No. NC4013681
Encompass_Preferred
YPD Broth, also written YEPD Broth, is the routine rich medium for growing Saccharomyces cerevisiae in molecular biology and yeast research. Peptone and yeast extract supply nitrogen, vitamins and growth factors while dextrose provides fermentable carbon, final pH 6.5, giving rapid, dependable growth for propagation, transformation, plasmid work and protein expression. A laboratory staple wherever budding yeast are cultured. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013681 Supplier HIMEDIA Supplier No. M1363500G
YPD Broth, also written YEPD Broth, is the routine rich medium for growing Saccharomyces cerevisiae in molecular biology and yeast research. Peptone and yeast extract supply nitrogen, vitamins and growth factors while dextrose provides fermentable carbon, final pH 6.5, giving rapid, dependable growth for propagation, transformation, plasmid work and protein expression. A laboratory staple wherever budding yeast are cultured. Dehydrated powder, store in a cool dry place. Packaged as 500 g.