Learn More
Crescent Chemical Co Inc YPD MEDIUM POWDER 50G FOR 1L
SDP
Catalog No. 50249154
Encompass_Preferred
YPD Medium, Powder 50 g/L A nutritious general growth medium for the propagation of yeast.20 g/l Tryptone10 g/l Yeast extract20 g/l DextroseFor making 1 L liquid medium suspend 50 g in 1 L distilled water and sterilize by autoclaving. Size - 500G Storage Co
Catalog No. 50-249-154 Supplier Crescent Chemical Co Inc Supplier No. SE48507.01
YPD Medium, Powder 50 g/L A nutritious general growth medium for the propagation of yeast.20 g/l Tryptone10 g/l Yeast extract20 g/l DextroseFor making 1 L liquid medium suspend 50 g in 1 L distilled water and sterilize by autoclaving. Size - 500G Storage Co