Learn More
Zymo Research Corporation YPD Plus, 100 ml
SDP
Catalog No. NC9815070
Encompass
Following treatment of yeast with lithium acetate and polyethylene glycol solutions, it is often necessary to add medium to allow for a period of outgrowth, which normally results in increased cell survival. This is useful when attempting to maximize transformation efficiencies for library screening or transforming yeast with multiple plasmids. YPD Plus is a special formulation of conventional YPD outgrowth medium that increases yeast transformation efficiencies by > 50%. YPD Plus is recommended for mutant yeast strains exhibiting poor growth characteristics that are not amenable to transformation. Simply supplement a yeast transformation reaction mixture with YPD Plus to achieve consistent increases in yeast transformation efficiencies.
Catalog No. NC9815070 Supplier Zymo Research Corporation Supplier No. Y1003100
Following treatment of yeast with lithium acetate and polyethylene glycol solutions, it is often necessary to add medium to allow for a period of outgrowth, which normally results in increased cell survival. This is useful when attempting to maximize transformation efficiencies for library screening or transforming yeast with multiple plasmids. YPD Plus is a special formulation of conventional YPD outgrowth medium that increases yeast transformation efficiencies by > 50%. YPD Plus is recommended for mutant yeast strains exhibiting poor growth characteristics that are not amenable to transformation. Simply supplement a yeast transformation reaction mixture with YPD Plus to achieve consistent increases in yeast transformation efficiencies.