Learn More
Aviva Systems Biology Corp YPEL5 Antibody - N-terminal region (ARP56822_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56822_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YPEL5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human YPEL5. Alternate names: CGI-127ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-217