Learn More
Teknova YPG Broth, Sterile. 8 pack.
SDP
Catalog No. 502387918
Encompass
2.00% Tryptone, 1.00% Yeast Extract, 2.00% D-(+)-Galactose
Catalog No. 50-238-7918 Supplier Teknova Supplier No. Y54108PK
2.00% Tryptone, 1.00% Yeast Extract, 2.00% D-(+)-Galactose