Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc YPG Agar Plates, Case 100 X 30 mL
SDP

Catalog No. 507517596
Encompass_Preferred

YPG Agar Plates is recommended for the growth of Saccharomyces cerevisiae for molecular biology purpose. YPG is a complex medium containing a nonfermentable carbon source (glycerol) that does not support the growth of ρ– or pet mutants.

Ready to use in 30 mL pertri dishes, 10 plates per sleeve
Re-closable zipper pouch for convenient storage

Applications: YPG Plates are useful for isolation of Beta Carotene-Hyper producing Mutants and biocatalyst preparation.

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Autoclave Sterilized

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7596 Supplier Quality Biological Inc Supplier No. 340190301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YPG Agar Plates is recommended for the growth of Saccharomyces cerevisiae for molecular biology purpose. YPG is a complex medium containing a nonfermentable carbon source (glycerol) that does not support the growth of ρ– or pet mutants.

Ready to use in 30 mL pertri dishes, 10 plates per sleeve
Re-closable zipper pouch for convenient storage

Applications: YPG Plates are useful for isolation of Beta Carotene-Hyper producing Mutants and biocatalyst preparation.

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Autoclave Sterilized

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.