Learn More
Medchemexpress LLC YQ128 | 2454246-18-3 | >99.7% | 100 MG
SDP
Catalog No. 5000338353
Encompass_Preferred
YQ128 is a potent and selective second-generation NLRP3 (NOD-like receptor P3) inflammasome inhibitor with an IC50 of 0.30 μM. It significantly and selectively suppresses the production of IL-1β, but not TNF-α. This compound can cross the blood-brain barrier (BBB) to reach the central nervous system (CNS) and has demonstrated anti-inflammatory activity. YQ128 is also a click chemistry reagent, featuring an Alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide-containing molecules. It is intended for research use only.
  • Potent and selective second-generation NLRP3 inflammasome inhibitor
  • Significantly and selectively suppresses IL-1β production
  • Crosses the blood-brain barrier to the central nervous system
  • Exhibits anti-inflammatory activity
  • Functions as a click chemistry reagent
Catalog No. 50-003-38353 Supplier Medchemexpress LLC Supplier No. HY130252100MG
YQ128 is a potent and selective second-generation NLRP3 (NOD-like receptor P3) inflammasome inhibitor with an IC50 of 0.30 μM. It significantly and selectively suppresses the production of IL-1β, but not TNF-α. This compound can cross the blood-brain barrier (BBB) to reach the central nervous system (CNS) and has demonstrated anti-inflammatory activity. YQ128 is also a click chemistry reagent, featuring an Alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide-containing molecules. It is intended for research use only.
  • Potent and selective second-generation NLRP3 inflammasome inhibitor
  • Significantly and selectively suppresses IL-1β production
  • Crosses the blood-brain barrier to the central nervous system
  • Exhibits anti-inflammatory activity
  • Functions as a click chemistry reagent