Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YQA14 10mg | 1221408-42-9 | 442.94 | C23H27ClN4O3 | 10 MG
SDP

Catalog No. 5000226421
Encompass_Preferred

YQA14 is a high-affinity dopamine D3 receptor antagonist used in preclinical research on addiction and reward. It reduces morphine- and cocaine-induced conditioned place preference in animal models and is supplied as a milligram-scale research compound for laboratory use.

  • High-affinity dopamine D3 receptor antagonist.
  • Reduces morphine- and cocaine-induced conditioned place preference in animals.
  • Chemical formula C23H27ClN4O3; molecular weight 442.94.
  • Reported purity 98.6% (HPLC).
  • Suitable for in vitro and in vivo pharmacology studies; handle as a research chemical.
  • Store at 4°C and protect from light; in solvent, store at -80°C for long-term.

Catalog No. 50-002-26421 Supplier Medchemexpress LLC Supplier No. HY11618110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YQA14 is a high-affinity dopamine D3 receptor antagonist used in preclinical research on addiction and reward. It reduces morphine- and cocaine-induced conditioned place preference in animal models and is supplied as a milligram-scale research compound for laboratory use.

  • High-affinity dopamine D3 receptor antagonist.
  • Reduces morphine- and cocaine-induced conditioned place preference in animals.
  • Chemical formula C23H27ClN4O3; molecular weight 442.94.
  • Reported purity 98.6% (HPLC).
  • Suitable for in vitro and in vivo pharmacology studies; handle as a research chemical.
  • Store at 4°C and protect from light; in solvent, store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.