Learn More
Cayman Chemical YS-035 (hydrochloride), Purity: ≥98%, CAS: 89805-39-0, 50 mg
SDP
Catalog No. 502966576
Encompass_Preferred
A verapamil derivative that dose-dependently blocks calcium uptake via Na+/Ca2+ exchange in diverse types of cells (Ki = 28 µM); also prevents calcium release from mitochondria induced by Ruthenium Red but not by uncouplers, respiratory inhibitors, or a chelating agent
Catalog No. 50-296-6576 Supplier Cayman Chemical Supplier No. 1770550 MG
A verapamil derivative that dose-dependently blocks calcium uptake via Na+/Ca2+ exchange in diverse types of cells (Ki = 28 µM); also prevents calcium release from mitochondria induced by Ruthenium Red but not by uncouplers, respiratory inhibitors, or a chelating agent