Learn More
Medchemexpress LLC YS-201 | 108852-42-2 | 457.52 | 20 MG
SDP
Catalog No. 5000447601
Encompass_Preferred
YS-201 is a dihydropyridine-type calcium channel antagonist. It has potential applications in the treatment of angina pectoris and hypertension. In vivo, YS-201 (Diperdipine) has been shown to reduce systemic vascular resistance and improve stroke index and left ventricular ejection fraction. It has an absolute bioavailability of 18.7% after intravenous and oral doses, with biliary excretion accounting for about 0.1% of its total clearance.
  • Dihydropyridine-type calcium channel antagonist
  • Potential for angina pectoris and hypertension treatment
  • Reduces systemic vascular resistance
  • Improves stroke index and left ventricular ejection fraction
  • Absolute bioavailability of 18.7% after intravenous and oral doses
  • Biliary excretion accounts for approximately 0.1% of total clearance
Catalog No. 50-004-47601 Supplier Medchemexpress LLC Supplier No. HYU0013720MG
YS-201 is a dihydropyridine-type calcium channel antagonist. It has potential applications in the treatment of angina pectoris and hypertension. In vivo, YS-201 (Diperdipine) has been shown to reduce systemic vascular resistance and improve stroke index and left ventricular ejection fraction. It has an absolute bioavailability of 18.7% after intravenous and oral doses, with biliary excretion accounting for about 0.1% of its total clearance.
  • Dihydropyridine-type calcium channel antagonist
  • Potential for angina pectoris and hypertension treatment
  • Reduces systemic vascular resistance
  • Improves stroke index and left ventricular ejection fraction
  • Absolute bioavailability of 18.7% after intravenous and oral doses
  • Biliary excretion accounts for approximately 0.1% of total clearance