Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YS-363 | 2470908-90-6 | MFCD12195663 | 99.5% | 494.58 | C30H30N4O3 | 10 MG
SDP

Catalog No. 5000286854
Encompass_Preferred

YS-363 is a potent, selective, orally active epidermal growth factor receptor (EGFR) inhibitor provided for research use. It inhibits wild-type and L858R mutant EGFR with IC50 values of 0.96 nM and 0.67 nM, respectively, and has been reported to induce G0/G1 cell cycle arrest and apoptosis. The compound is supplied as a light yellow solid and as DMSO solutions for experimental flexibility.

  • Potent EGFR inhibitor with sub-nanomolar to low-nanomolar IC50 values.
  • Induces G0/G1 cell cycle arrest and apoptosis in cellular assays.
  • Molecular formula: C30H30N4O3.
  • Molecular weight: 494.58 g/mol.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats for flexible use.

Catalog No. 50-002-86854 Supplier Medchemexpress LLC Supplier No. HY15528510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YS-363 is a potent, selective, orally active epidermal growth factor receptor (EGFR) inhibitor provided for research use. It inhibits wild-type and L858R mutant EGFR with IC50 values of 0.96 nM and 0.67 nM, respectively, and has been reported to induce G0/G1 cell cycle arrest and apoptosis. The compound is supplied as a light yellow solid and as DMSO solutions for experimental flexibility.

  • Potent EGFR inhibitor with sub-nanomolar to low-nanomolar IC50 values.
  • Induces G0/G1 cell cycle arrest and apoptosis in cellular assays.
  • Molecular formula: C30H30N4O3.
  • Molecular weight: 494.58 g/mol.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats for flexible use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.