Learn More
Medchemexpress LLC YS-370 | 2470908-79-1 | 98.1% | C37H35BrN4O3 | 10MG
SDP
Catalog No. 5000261707
Encompass_Preferred
YS-370 is a small-molecule, orally active inhibitor of P-glycoprotein used in transporter and pharmacokinetic research. It stimulates P-gp ATPase activity and has been characterized for potency and selectivity, making it suitable for mechanistic studies and in vivo evaluation.
  • Potent and selective inhibitor of P-glycoprotein.
  • Orally active small molecule appropriate for in vivo studies.
  • Stimulates P-gp ATPase activity for mechanistic assays.
  • High purity (98.1%) suitable for research applications.
  • Solid, white to off-white powder with DMSO solubility of 8.33 mg/mL.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-61707 Supplier Medchemexpress LLC Supplier No. HY13286610MG
YS-370 is a small-molecule, orally active inhibitor of P-glycoprotein used in transporter and pharmacokinetic research. It stimulates P-gp ATPase activity and has been characterized for potency and selectivity, making it suitable for mechanistic studies and in vivo evaluation.
  • Potent and selective inhibitor of P-glycoprotein.
  • Orally active small molecule appropriate for in vivo studies.
  • Stimulates P-gp ATPase activity for mechanistic assays.
  • High purity (98.1%) suitable for research applications.
  • Solid, white to off-white powder with DMSO solubility of 8.33 mg/mL.
  • Stable under recommended storage conditions for long-term use.