Learn More
Medchemexpress LLC Ys-370 | 2470908-79-1 | MFCD34676892 | 98.1% | 663.60 g/mol | C37H35BrN4O3 | 5 MG
SDP
Catalog No. 5000262238
Encompass_Preferred
YS-370 is a small-molecule P-glycoprotein (P-gp) inhibitor for research use. It is a white to off-white solid (CAS 2470908-79-1) with molecular formula C37H35BrN4O3 and molecular weight 663.60 g/mol, intended for in vitro and preclinical P-gp inhibition studies.
  • High purity, 98.1% (reported by HPLC).
  • Solid form, white to off-white, stable for storage.
  • Available in small pack sizes for screening, 5 mg to 100 mg.
  • Suitable for in vitro and in vivo P-gp inhibition assays.
  • Analytical data available: COA, H NMR, LC-MS.
Catalog No. 50-002-62238 Supplier Medchemexpress LLC Supplier No. HY1328665MG
YS-370 is a small-molecule P-glycoprotein (P-gp) inhibitor for research use. It is a white to off-white solid (CAS 2470908-79-1) with molecular formula C37H35BrN4O3 and molecular weight 663.60 g/mol, intended for in vitro and preclinical P-gp inhibition studies.
  • High purity, 98.1% (reported by HPLC).
  • Solid form, white to off-white, stable for storage.
  • Available in small pack sizes for screening, 5 mg to 100 mg.
  • Suitable for in vitro and in vivo P-gp inhibition assays.
  • Analytical data available: COA, H NMR, LC-MS.