Learn More
TARGETMOL CHEMICALS INC YS-49 10MG
SDP
Catalog No. 502581352
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YS-49 is an activator of PI3K/Akt (a downstream target of RhoA). purity: 99%
Catalog No. 50-258-1352 Supplier TARGETMOL CHEMICALS INC Supplier No. T1337610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YS-49 is an activator of PI3K/Akt (a downstream target of RhoA). purity: 99%