Learn More
Selleck Chemical LLC YS-49-E0785-5MG
SDP
Catalog No. 502389869
Encompass_Preferred
YS-49 is a PI3K/Akt (a downstream target of RhoA) activator to reduce RhoA/PTEN activation in the 3-methylcholanthrene-treated cells inhibits angiotensin II (Ang II)-stimulated proliferation of VSMCs via induction of heme oxygenase (HO)-1 also is an isoquinoline compound alkaloid has a strong positive inotropic action through activation of cardiac -adrenoceptors
Catalog No. 50-238-9869 Supplier Selleck Chemical LLC Supplier No. E07855MG
YS-49 is a PI3K/Akt (a downstream target of RhoA) activator to reduce RhoA/PTEN activation in the 3-methylcholanthrene-treated cells inhibits angiotensin II (Ang II)-stimulated proliferation of VSMCs via induction of heme oxygenase (HO)-1 also is an isoquinoline compound alkaloid has a strong positive inotropic action through activation of cardiac -adrenoceptors