Learn More
Medchemexpress LLC YS-49 monohydrate | 3028631-24-2 | C20H22BrNO3 | 5 MG
SDP
Catalog No. 5000376934
Encompass_Preferred
YS-49 monohydrate is a PI3K/Akt activator that reduces RhoA/PTEN activation in 3-methylcholanthrene-treated cells. It inhibits angiotensin II (Ang II)-stimulated proliferation of VSMCs via induction of heme oxygenase (HO)-1 and acts as an isoquinoline compound alkaloid with strong positive inotropic action through activation of cardiac β-adrenoceptors.
  • Activates PI3K/Akt pathway
  • Reduces RhoA/PTEN activation
  • Inhibits angiotensin II-stimulated proliferation of VSMCs
  • Induces heme oxygenase (HO)-1
  • Exhibits strong positive inotropic action via cardiac β-adrenoceptor activation
Catalog No. 50-003-76934 Supplier Medchemexpress LLC Supplier No. HY15477A5MG
YS-49 monohydrate is a PI3K/Akt activator that reduces RhoA/PTEN activation in 3-methylcholanthrene-treated cells. It inhibits angiotensin II (Ang II)-stimulated proliferation of VSMCs via induction of heme oxygenase (HO)-1 and acts as an isoquinoline compound alkaloid with strong positive inotropic action through activation of cardiac β-adrenoceptors.
  • Activates PI3K/Akt pathway
  • Reduces RhoA/PTEN activation
  • Inhibits angiotensin II-stimulated proliferation of VSMCs
  • Induces heme oxygenase (HO)-1
  • Exhibits strong positive inotropic action via cardiac β-adrenoceptor activation