Learn More
Medchemexpress LLC Ys-67 | 2761327-15-3 | 99.4% | 522.64 | C32H34N4O3 | 10 MG
SDP
Catalog No. 5000288227
Encompass_Preferred
YS-67 is a small-molecule epidermal growth factor receptor (EGFR) inhibitor for research use. It potently inhibits EGFR (IC50 = 5.2 nM), reduces p-EGFR and p-AKT signaling, and inhibits proliferation of A549, PC-9, and A431 cell lines.
  • Potent EGFR inhibition (IC50 = 5.2 nM).
  • Reduces phosphorylation of EGFR and AKT in cellular assays.
  • Inhibits proliferation of A549, PC-9, and A431 cell lines.
  • High purity (99.4%) suitable for in vitro studies.
  • Molecular formula C32H34N4O3; molecular weight 522.64.
  • Available in multiple small-scale quantities for laboratory testing.
Catalog No. 50-002-88227 Supplier Medchemexpress LLC Supplier No. HY15741310MG
YS-67 is a small-molecule epidermal growth factor receptor (EGFR) inhibitor for research use. It potently inhibits EGFR (IC50 = 5.2 nM), reduces p-EGFR and p-AKT signaling, and inhibits proliferation of A549, PC-9, and A431 cell lines.
  • Potent EGFR inhibition (IC50 = 5.2 nM).
  • Reduces phosphorylation of EGFR and AKT in cellular assays.
  • Inhibits proliferation of A549, PC-9, and A431 cell lines.
  • High purity (99.4%) suitable for in vitro studies.
  • Molecular formula C32H34N4O3; molecular weight 522.64.
  • Available in multiple small-scale quantities for laboratory testing.