Learn More
YSI™ 2002 Pro Series Replacement Galvanic DO Sensor
Catalog No. 15177458 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
For Use With (Application):
Dissolved Oxygen testing
Catalog No. For Use With (Application)
15-177-458 Dissolved Oxygen testing
1 options
Catalog No. 15-177-458 Supplier YSI™ Supplier No. 605202
Add to Cart
Add to Cart
Includes
Sensor, six membrane caps and bottle of solution
Requires
Cables must be ordered separately
User-replaceable dissolved oxygen sensor
For use in dissolved oxygen testing with YSI Professional Plus and Pro20 instruments.
  • Easy replacement with no tools necessary
  • Convenient screw-on cap membranes
Warranty and Services
Six months
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Professional Plus and Pro20 instruments
For Use With (Application) Dissolved Oxygen testing
Warranty 6 Months
Probe Galvanic DO
Includes YSI 2002 DO Sensor, 6 membrane caps, bottle of solution