Learn More
YSI™ 6051020 Pro Series pH/ORP/ISE and DO Field Cable
Catalog No. 15177650 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Length (Metric) Cable:
30 m
Catalog No. Length (Metric) Cable
15-177-650 30 m
1 options
Catalog No. 15-177-650 Supplier YSI™ Supplier No. 605102030
Add to Cart
Add to Cart
For use with Pro Plus and Pro1020 instruments.
  • 2-port cable
  • One sensor port accepts a pH, ORP, or ISE sensor while the second port accepts a dissolved oxygen (DO) sensor
  • Cable Length: 30m
  • Includes a cable management kit and integrated temperature sensor
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Pro Plus and Pro1020 Instruments
Includes Cable management kit and integrated temperature sensor
Length (Metric) Cable 30 m