Learn More
Description
- Contains two bottles each of pH 4, 7, and 10 calibration buffers (Cat. Nos. 3821, 3822, and 3823)
- Plastic bottles (475mL/ea.)
Expiration Dates
- Unopended: 2 years
- Opened: 6 months
Specifications
Specifications
| For Use With (Application) | pH Calibration |
| For Use With (Equipment) | pH Meter |
| pH | 4, 7, 10 |
| Physical Form | Liquid |
| Quantity | 1 Kit (2 bottles ea. 3821,3822 and 3823) |
| Solution Type | pH Buffer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.