Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Confidence Solution, YSI™

Catalog No. 15176216 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-176-216 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-176-216 Supplier Yellow Springs Instrument (YSI) Supplier No. 005580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Verification check only for pH, ORP, conductivity—not considered a calibration solution

  • This solution is designed for verifying measurements, not calibration
  • Simply immerse multiparameter probes in the solution and verify the pH, ORP, and conductivity values instantly
TRUSTED_SUSTAINABILITY

Specifications

Packaging 475mL
Analyte or Component Names Confidence Solution
Element Confidence Solution
Quantity 475 mL
Chemical Name or Material Confidence Solution
ShelfLife Unopened: 2 years, Opened: 3 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.