Learn More
Cole-Parmer YSI 5908 Replacement DO Cap Membrane Kit; 6 Yellow Screw-on Cap Membranes (1.25 mil PE), Bottle of Electrolyte, and Sanding Disc
Catalog No. NC1790761 Shop All Cole-Parmer Products
Encompass
Six yellow screw-on cap membranes (1.25 mil PE), a bottle of electrolyte, and 1 sanding disc. Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.
Catalog No. NC1790761 Supplier Cole-Parmer Supplier No. 9985280
Six yellow screw-on cap membranes (1.25 mil PE), a bottle of electrolyte, and 1 sanding disc. Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.