Learn More
YSI™ Electrode Replacement Kit for EcoSense™ EC30A Instrument
Catalog No. 15177851 Shop All Yellow Springs Instrument (YSI) Products
Catalog No. 15-177-851 Supplier YSI™ Supplier No. 606114
Add to Cart
Add to Cart
Includes
Electrode, electrode collar, two o-rings and sensor cap
Replacement electrode kit for EcoSense EC30A Conductivity/TDS/Temperature Pen.
For use with EcoSense EC30A Conductivity/TDS/Temperature pen-style instrument.
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode Replacement Kit
Includes Electrode, electrode collar, two o-rings, and sensor cap