Learn More
YSI™ Flow Cell Kit, 1- or 2-Port Sensor for Pro Plus
Catalog No. 15178108 Shop All Yellow Springs Instrument (YSI) Products
Catalog No. 15-178-108 Supplier YSI™ Supplier No. 603077
Add to Cart
Add to Cart
Includes
Flow Cell, Standard, 203mL (603059) and Flow Cell Adapter, Single Port (603078)
Can be used with all 1 or 2 port Professional Series probe assembly.
Also can be used with 550, 550A, 60 meters.
TRUSTED_SUSTAINABILITY
Specifications
Type Flow Cell Kit
Includes Flow Cell, Standard, 203mL (603059) and Flow Cell Adapter, Single Port (603078)