Learn More
YSI™ Model 5739 Field Oxygen/Temperature Probe
Catalog No. 1329943 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
For Use With (Application):
Calibrated easily and quickly
Catalog No. For Use With (Application)
13-299-43 Calibrated easily and quickly
1 options
Catalog No. 13-299-43 Supplier YSI™ Supplier No. 057087
Includes
One 13-299-16 Membrane/KCI Kit
Requires
Detachable cable; order separately.
Can be calibrated easily and quickly
For use with YSI portable Models 52 and 58 or with YSI 5000 or 5100 benchtop meters when equipped with optional Probe Adapter Cable 13-299-78.
  • Calibrates easily in air in just seconds
  • Automatic temperature compensation and pressure compensation to ±0.5% of reading at 100psi/230 ft. (7kg/cm2/70m) of water
  • Rugged plastic body, built-in sinker weight
Benchtop meters are not recommended for field use. Not for use with handheld Models 55, 85, 95, 550 or 556.
Order Info
One 13-299-16 Membrane/KCI Kit
TRUSTED_SUSTAINABILITY
Specifications
Response Time 90% in 30 sec.
Temperature (English) Operating 0°F to 113°F
Temperature (Metric) Range 0°C to 45°C
Includes Membrane/KCI kit
Warranty 1 Year
For Use With (Equipment) YSI portable model 52 and 58 or YSI 5000 and 5100 benchtop meter
Probe Type Field/O2 Temperature
For Use With (Application) Calibrated easily and quickly
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.