Learn More
YSI™ BNC Adapter for MultiLab Line Benchtop Laboratory Instrument
Catalog No. 09324572 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
For Use With (Equipment):
MultiLab 4010-2 Instruments, MultiLab 4010-3 Instruments
Catalog No. For Use With (Equipment)
09-324-572 MultiLab 4010-2 Instruments, MultiLab 4010-3 Instruments
1 options
Catalog No. 09-324-572 Supplier YSI™ Supplier No. 108132Y
Add to Cart
Add to Cart
For use with intelligent digital sensors and laboratory instruments.
  • For use with MultiLab 4010-2 or 4010-3 Instruments
  • For pH, ORP, and ISE electrodes with BNC connection
  • Converts one digital channel on MultiLab 4010-2/3 instruments to an analog signal
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MultiLab 4010-2 Instruments, MultiLab 4010-3 Instruments