Learn More
YSI™ MultiLab Line BOD Laboratory Kits
Catalog No. 09324565 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Channels:
1
Catalog No. Channels
09-324-565 1
1 options
Catalog No. 09-324-565 Supplier YSI™ Supplier No. 626570
Add to Cart
Add to Cart
Request a Quote
Kit for BOD calculation
BOD Lab Kit Includes
  • 4010-1 One Channel benchtop instrument
  • Optical BOD probe
Advanced BOD Lab Kit Includes
  • 4010-1 One Channel benchtop instrument
  • Optical BOD probe
  • BOD Analyst Pro desktop software
TRUSTED_SUSTAINABILITY
Specifications
Channels 1
Includes 4010-1 One channel benchtop instrument, optical BOD probe