Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ ProPlus Polarographic DO/Conductivity Kit with 10m Cable

Catalog No. 09324530 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Backlight Option:
Yes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Backlight Option
09-324-530 Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-324-530 Supplier YSI™ Supplier No. 603182

Includes

Pro Plus handheld, 10m cable with polarographic DO and conductivity probe, 5908 cap membrane kit (1.25 mil PE), soft-sided carrying case

Includes everything from lab to field along with polarographic DO and conductivity probe.

Pro Plus Meters

  • Rubber, over-molded IP67 waterproof case
  • Rugged cables, sensors, and military-spec connectors
  • Drop-tested 1m at all angles
  • Scratch-resistant lens; viewable with polarized glasses
  • Instrument floats
  • Backlit display and keypad
  • 50 data-set memory ensures data integrity
  • USB connection powers the instrument
  • Auto-buffer recognition; up to 6-point cal
  • Detailed GLP data
  • Power: 2 alkaline C-cells provide 400 continuous hours at ambient temperature without backlight
  • Free, Data Manager desktop software and ProComm II saddle

Certifications

CE, WEEE

Compliance

RoHS

Warranty and Services

3-year instrument warranty; 2-year cable warranty

TRUSTED_SUSTAINABILITY

Specifications

Backlight Option Yes
Battery Life 400 continuous hours at ambient temperature without backlight
Battery Type 2 C Alkaline
Calibration Points Up to 6
Certifications/Compliance RoHS, CE, WEEE
Data Management Details Free, Data Manager desktop software and ProComm II saddle
Display Type Backlit
IP Rating IP67
Includes Pro Plus handheld, 10m cable with polarographic DO and conductivity probe, 5908 cap membrane kit (1.25 mil PE), soft-sided carrying case
Keypad Backlit
Memory 50 data sets
Power Supply 2 alkaline C-cells
Product Type ProPlus Polarographic DO/Conductivity Kit with 10m Cable
Warranty 3-year instrument warranty; 2-year cable warranty
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.