Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ ProPlus Galvanic DO/pH/ORP/Conductivity Quatro Kit with 4m Cable

Catalog No. 09324541 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Backlight Option:
Yes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Backlight Option
09-324-541 Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-324-541 Supplier YSI™ Supplier No. 603224

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Includes

Pro Plus handheld, 4m Quatro cable with galvanic DO, pH/ORP and conductivity sensors, 5913 cap membrane kit (1.25 mil PE), soft-sided carrying case

Includes everything from lab to field along with galvanic DO, pH/ORP and conductivity sensors.

Pro Plus Meters

  • Rubber, over-molded IP67 waterproof case
  • Rugged cables, sensors, and military-spec connectors
  • Drop-tested 1m at all angles
  • Scratch-resistant lens; viewable with polarized glasses
  • Instrument floats
  • Backlit display and keypad
  • 50 data-set memory ensures data integrity
  • USB connection powers the instrument
  • Auto-buffer recognition; up to 6-point cal
  • Detailed GLP data
  • Power: 2 alkaline C-cells provide 400 continuous hours at ambient temperature without backlight
  • Free, Data Manager desktop software and ProComm II saddle

Certifications

CE, WEEE

Compliance

RoHS

Warranty and Services

3-year instrument warranty; 2-year cable warranty

TRUSTED_SUSTAINABILITY

Specifications

Includes Pro Plus handheld, 4m Quatro cable with galvanic DO, pH/ORP and conductivity sensors, 5913 cap membrane kit (1.25 mil PE), soft-sided carrying case
Calibration Up to 6-point cal
Display Type Backlit
Backlight Option Yes
Memory 50 data-sets
Data Management Details Free, Data Manager desktop software and ProComm II saddle
Certifications/Compliance RoHS, CE, WEEE
IP Rating IP67
Warranty 3 Year Instrument, 2 Year Cable
Model ProPlus
Power Supply 2 alkaline C-cells
Keypad Backlit
Type ProPlus Galvanic DO/pH/ORP/Conductivity Quatro Kit
Battery Type 2 Alkaline C-cells
Battery Life 400 continuous hr. at ambient temperature without backlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.