Learn More
YSI™ ProPlus Polarographic DO/pH/ORP/Conductivity Quatro Kit w/4m Cable
Shop All Yellow Springs Instrument (YSI) ProductsDescription
Includes
Pro Plus handheld, 4m Quatro cable with polarographic DO, pH/ORP and conductivity sensors, 5908 cap membrane kit (1.25 mil PE), soft-sided carrying case
Pro Plus Meters
- Rubber, over-molded IP67 waterproof case
- Rugged cables, sensors, and military-spec connectors
- Drop-tested 1m at all angles
- Scratch-resistant lens; viewable with polarized glasses
- Instrument floats
- Backlit display and keypad
- 50 data-set memory ensures data integrity
- USB connection powers the instrument
- Auto-buffer recognition; up to 6-point cal
- Detailed GLP data
- Power: 2 alkaline C-cells provide 400 continuous hours at ambient temperature without backlight
- Free, Data Manager desktop software and ProComm II saddle
Certifications
CE, WEEE
Compliance
RoHS
Warranty and Services
3-year instrument warranty; 2-year cable warranty
Specifications
Specifications
| Backlight Option | Yes |
| Battery Life | 400 continuous hours at ambient temperature without backlight |
| Battery Type | 2 C Alkaline |
| Calibration Points | Up to 6 |
| Certifications/Compliance | RoHS, CE, WEEE |
| Data Management Details | Free, Data Manager desktop software and ProComm II saddle |
| Display Type | Backlit |
| IP Rating | IP67 |
| Includes | Pro Plus handheld, 4m Quatro cable with polarographic DO, pH/ORP and conductivity sensors, 5908 cap membrane kit (1.25 mil PE), soft-sided carrying case |
| Keypad | Backlit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.