Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Science pH Combination Electrode with Silamid™ Double Junction, pHT-G

Catalog No. 15177841 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
pH Range:
0 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
15-177-841 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-177-841 Supplier YSI™ Supplier No. 400361
Add to Cart
Add to Cart

Designed to meet the requirements of a wide variety of laboratory applications.

The Science pHT-G features a ground joint junction with a relatively fast outflow of electrolyte (approx.3mL per day). This fast outflow helps keep the junction clear, which makes the Science pHT-G electrode suitable for measurements in solutions with a lot suspended particles.

  • 3-in-1 double ground joint junction
  • For use with TruLab Instruments
  • Glass shaft
  • Measures pH and temperature
  • BNC connector and one banana plug
  • Buffer recognition
  • User calibratable and replaceable
  • Refillable
  • Sphere (bulb) membrane
  • Reference Electrolyte: 3M KCl liquid

Specifications

  • pH Range: 0 to 14
  • Temperature Range: -5° to +100°C (23° to 212°F)
  • NTC: 30 kOhm
  • Cable Length: 1m
  • Length: 170mm
  • Dia.: 12mm
  • Min. Immersion Depth: 12 to 14mm

Order Info

Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time.

Compliance

GLP compliant

Warranty and Services

One year

TRUSTED_SUSTAINABILITY

Specifications

Type Science pHT-G pH Combination Electrode
For Use With (Application) Measuring pH and temperature
Includes 1m cable, BNC connector, one banana plug
Material Glass
pH Range 0 to 14
Temperature (Metric) Range -5°C to +100°C
For Use With (Equipment) TruLab Instruments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.