Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ TruLab™ pH 1310P pH/mV Single Channel Laboratory Benchtop Meter

Catalog No. 15177828 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Range (pH):
-2.0-20.0, -2.00-20.00, -2.000-19.999
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
15-177-828 -2.0-20.0, -2.00-20.00, -2.000-19.999
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-177-828 Supplier YSI™ Supplier No. 1AA320PY
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Meter, universal AC power supply, electrode stand, manual, software and USB cable

Easy-to-use meter to accurately measure pH, mV (ORP) and temperature in lab with integrated thermal printer

  • Single channel measurements
  • AutoRead function improves repeatability
  • Easy-to-read backlit display with text menu for easy operation for pH, mV (ORP) and temperature
  • Fast data transfer via USB in .csv or ASCII format or print with integrated thermal printer
  • Easy calibration with adjustable calibration timer, which indicates when to perform the next calibration routine
  • Automatic or manual temperature compensation
  • 1, 2, 3, 4 or 5 point calibration for pH; 22 stored buffer sets for auto-buffer recognition and easy calibration
  • BNC connection for pH, mV; banana connection for temperature probes
  • Continuous Monitoring Control (CMC) function for monitoring the ideal measuring range for pH measurements based on the last calibration
  • Wide selection of electrodes: TruLab analog pH/ORP/temperature probes (sold separately)
  • Four alkaline AA batteries provides approx.150 hours; universal power supply; also compatible with four NiMH AA rechargeables (instrument does not charge rechargeable batteries)

Specifications:

  • Buffer Recognition: Yes
  • Sampling: Yes
  • User Calibratable: Yes
  • Logging Capabilities: Yes
  • Operating pH Range: -2.0 to 20.0; -2.00 to 20.00; -2.000 to 19.999
  • pH Resolution: 0.1, 0.01, 0.001
  • pH Accuracy (±1 digit): ±0.1, ±0.01, ±0.005 (temp. of test sample 15° to 35°C; within a range of ±2 pH of the calibration point)
  • mV Range: -1200 to 1200.0; -2500 to 2500
  • mV Resolution: 0.1, 1.0
  • mV Accuracy: ±0.3, ±1.0 (temp of test sample 15° to 35°C)
  • Operating Temperature Range: -5° to +105°C (23° to 221°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.1
  • Memory: 5000 data sets auto logging, 500 data sets manual logging
  • Manual Temperature Input: -25° to +130°C (-13° to +266°F) in increments of 1
  • Storage Temperature: -25° to +65°C (-13° to +149°F)
  • Dimensions (L x W x H): 28 x 23 x 8cm (11 x 9 x 3.2 in.)
  • Weight: 1kg (2 lb.)

Order Info

Cables, probes/sensors and accessories sold separately.

Certifications

cETLus, IP43, CE, electrical protective class III

Compliance

GLP/AQA compliant

Warranty and Services

Three years

TRUSTED_SUSTAINABILITY

Specifications

Description pH 1310P pH/mV Single Channel Laboratory Benchtop Meter
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.3, ±1.0
Accuracy (pH) ±0.1, ±0.01, ±0.005
Data Management Details USB or Integrated Thermal Printer
Includes Meter, universal AC power supply, electrode stand, manual, software and USB cable
Range (pH) -2.0-20.0, -2.00-20.00, -2.000-19.999
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) -1200 to 1200.0, -2500 to 2500
Resolution (mV) 0.1, 1.0
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 23°F to 221°F
Resolution (Temperature) 0.1
Display Type Digital
Data Points 5000 (Auto Logging), 500 (Manual Logging)
Inputs Single channel
Certifications/Compliance CE, cETLus; electrical protective class III; IP43
Dimensions (L x W x H) 28 x 23 x 8 cm
Weight (Metric) 1 kg
Warranty 3 Years
Probe Type TruLab analog pH/ORP/temperature probes
Electrical Requirements Four alkaline AA batteries provides approx.150 hours; universal power supply; also compatible with four NiMH AA rechargeables (instrument does not charge rechargeable batteries)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.