Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Truline Lab Selective Ion Electrode, Calcium (Ca++)

Catalog No. 09324577 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Parameter:
Calcium (CA++)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Parameter
09-324-577 Calcium (CA++)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-324-577 Supplier YSI™ Supplier No. 400387

missing translation for 'validMainframeMsg'

Uses four-sensor technology

  • ISE Combination electrode
  • Calcium (Ca++)
  • Polymer/PVC membrane; Double Junction
  • 0.02 to 40,000mg/L 
  • pH: 2.5 to 11
  • Use for aquariums, industrial, food/beverage applications
TRUSTED_SUSTAINABILITY

Specifications

Applications Aquariums, industrial, food/beverage
Filling Solution Type Reference/Electrode Filling
Internal Reference Double Junction
Operating Range (Metric) 0.02 to 40,000mg/L Ca2+
pH Range 2.5 to 11
Interferences Pb2+, Hg2+, Si2+, Mg2+, Cu2+, K+, Tris+, Na+, others
Electrode Polymer/PVC membrane
Parameter Calcium (CA++)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.