Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Truline Lab Selective Ion Electrode, Chloride (Cl-)

Catalog No. 09324578 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Parameter:
Chloride (Cl-)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Parameter
09-324-578 Chloride (Cl-)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-324-578 Supplier YSI™ Supplier No. 400393
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses four-sensor technology

  • ISE Combination electrode
  • Chloride (Cl-)
  • Solid state; Double Junction
  • 1.8 to 35,500mg/L 
  • pH: 2 to 12 
  • Use for industrial, wastewater, drinking water, food/beverage, environmental applications
  • Store in dark when not in use
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial, wastewater, drinking water, food/beverage, environmental. Store in dark when not in use.
Filling Solution Type Reference/Electrode Filling
Internal Reference Double Junction
Operating Range (Metric) 1.8 to 35,500mg/L Cl-
pH Range 2 to 12
Interferences CN-, Br-, I-, OH-, S2-, NH3
Electrode Solid state
Parameter Chloride (Cl-)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.