Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Truline Lab Selective Ion Electrode, Silver/Sulfide (Ag+/S2-)

Catalog No. 09324586 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Parameter:
Silver/Sulfide (Ag+/S2-)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Parameter
09-324-586 Silver/Sulfide (Ag+/S2-)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-324-586 Supplier YSI™ Supplier No. 400433

missing translation for 'validMainframeMsg'

Uses four-sensor technology

  • ISE Combination electrode
  • Silver/Sulfide (Ag+/S2-)
  • Solid state, Double Junction
  • 0.01 to 107,900mg/L (Silver); 0.003 to 32,100mg/L (Sulfide)
  • pH: 2 to 12
  • Use for industrial, environmental, food/beverage applications
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial, environmental, food/beverage
Filling Solution Type Reference/Electrode Filling
Internal Reference Double Junction
Operating Range (Metric) 0.01 to 107,900mg/L (Ag+); 0.003 to 32,100mg/L (S-)
pH Range 2 to 12
Interferences Hg2+
Electrode Solid state
Parameter Silver/Sulfide (Ag+/S2-)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.