Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ TruLine ORP Electrode with Gel Reference Electrolyte, ORP 51

Catalog No. 15177844 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Temperature (Metric) Range:
-5°C to +80°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature (Metric) Range
15-177-844 -5°C to +80°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-177-844 Supplier YSI™ Supplier No. 400356
Add to Cart
Add to Cart

Includes

BNC plug head connector, 1m cable

Single fiber junction benchtop electrode with gel reference electrolyte.

  • For use with TruLab Instruments
  • Measures ORP and mV
  • Epoxy shaft
  • Buffer recognition
  • User calibratable and user replaceable

Specifications

  • Operating Temperature: -5° to +80°C (23° to 176°F)

Order Info

Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time.

Compliance

GLP compliant

Warranty and Services

One year

TRUSTED_SUSTAINABILITY

Specifications

Type TruLine ORP 51 Combination Electrode
For Use With (Application) Measuring ORP and mV
Includes 1m cable, BNC plug head connector
Material Epoxy
Temperature (Metric) Range -5°C to +80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.