Learn More
YSI™ TruLine pH Combination Electrode with Polymer Reference Electrolyte, pH 27
Shop All Yellow Springs Instrument (YSI) ProductsDescription
Includes
BNC connector, 1m cable
- For use with TruLab Instruments
- Measures pH and Redox
- Glass shaft
- Plug head connector
- Buffer recognition
- User calibratable and user replaceable
Specifications
- pH Range: 0 to 13
- Operating Temperature: -5° to +50°C (23° to 122°F)
Order Info
Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time.
Compliance
GLP compliant
Warranty and Services
One year
Specifications
Specifications
| Type | TruLine pH 27 Combination Electrode |
| For Use With (Application) | Measuring pH and redox |
| Includes | 1m cable, BNC plug head connector |
| Material | Glass |
| pH Range | 0 to 13 |
| Temperature (Metric) Range | -5°C to +50°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.