Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ TruLine pH Combination Electrode with Refillable Liquid Reference System, pH 17

Catalog No. 15177833 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
pH Range:
0 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
15-177-833 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-177-833 Supplier YSI™ Supplier No. 400351
Add to Cart
Add to Cart

Includes

BNC connector, 1m cable

Platinum single junction, combination benchtop electrode.

  • For use with TruLab Instruments
  • Glass shaft
  • Buffer recognition
  • User calibratable and user replaceable
  • Refillable reference electrolyte: KCl 3 mol/L, Ag+ free

Specifications

  • pH Range: 0 to 14
  • Operating Temperature: -5° to 100°C (23° to 212°F)

Order Info

Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time.

Compliance

GLP Compliant

Warranty and Services

One year

TRUSTED_SUSTAINABILITY

Specifications

Type TruLine pH 17 Combination Electrode
For Use With (Application) Measuring pH and temperature
Includes 1m cable, BNC connector
Material Glass
pH Range 0 to 14
Temperature (Metric) Range -5°C to +100°C
For Use With (Equipment) TruLab Instruments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.