Learn More
TARGETMOL CHEMICALS INC YT 146 1MG
SDP
Catalog No. 502878718
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. YT 146 is a selective adenosine receptor A2 agonist with cardioprotective and vasodilatory effects that inhibits neointimal thickening after endothelial injury in rat femoral arteries. Purity 99.72%
Catalog No. 50-287-8718 Supplier TARGETMOL CHEMICALS INC Supplier No. T352821MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. YT 146 is a selective adenosine receptor A2 agonist with cardioprotective and vasodilatory effects that inhibits neointimal thickening after endothelial injury in rat femoral arteries. Purity 99.72%