Learn More
Bioworld YT broth (2x), 500 g
Catalog No. 50254403
Encompass_Preferred
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.Contains (g/L): Tryptone: 16 g Yeast Extract 10g Sodium Chloride 5gInstruction to Use: Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final ph@ 25°C: 6.8-7.2. This product is sold for laboratory research use only.
Catalog No. 50-254-403 Supplier Bioworld Supplier No. 306201032
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.Contains (g/L): Tryptone: 16 g Yeast Extract 10g Sodium Chloride 5gInstruction to Use: Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final ph@ 25°C: 6.8-7.2. This product is sold for laboratory research use only.