Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc YT Medium (2X), 1000 mL
SDP

Catalog No. 507517466
Encompass_Preferred

Ready-to-use growth medium Nutritionally rich for the growth of recombinant strains of E. coli. Suitable for non-selective cultivation of M13 bacteriophage and E. coli strains for cloning, DNA plasmid production and production of recombinant proteins, as well as selective cultivation when appropriate antibiotics are added. Tryptone and yeast extracts are found in large quantities within this medium Provides necessary nitrogen and growth factors that the bacteriophage need to replicate more rapidly and efficiently without depleting the host cells of these critical nutrients.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 24 months from date of

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7466 Supplier Quality Biological Inc Supplier No. 340023131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use growth medium Nutritionally rich for the growth of recombinant strains of E. coli. Suitable for non-selective cultivation of M13 bacteriophage and E. coli strains for cloning, DNA plasmid production and production of recombinant proteins, as well as selective cultivation when appropriate antibiotics are added. Tryptone and yeast extracts are found in large quantities within this medium Provides necessary nitrogen and growth factors that the bacteriophage need to replicate more rapidly and efficiently without depleting the host cells of these critical nutrients.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 24 months from date of

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.