Learn More
Crescent Chemical Co Inc YT MEDIUM POWDER 31G FOR 1L
SDP
Catalog No. 50249151
Encompass_Preferred
YT Medium, Powder 31 g/L For cultivation of E. coli and M13 bacteriophages in molecular biology.16 g/l Tryptone10 g/l Yeast extract5 g/l NaClFor making 1 L 2x liquid medium suspend 31 g in 900 ml distilled water, adjust the pH to 7.0 with approximately 0.2 ml of 5
Catalog No. 50-249-151 Supplier Crescent Chemical Co Inc Supplier No. SE48503.01
YT Medium, Powder 31 g/L For cultivation of E. coli and M13 bacteriophages in molecular biology.16 g/l Tryptone10 g/l Yeast extract5 g/l NaClFor making 1 L 2x liquid medium suspend 31 g in 900 ml distilled water, adjust the pH to 7.0 with approximately 0.2 ml of 5