Learn More
Bioworld YT Medium, 1 kg
Catalog No. 50254409
Encompass_Preferred
Used for the cultivation of recombinant strains of Escherichia coli. Composition per liter:Bacto Tryptone, 8gBacto Yeast Extract, 5g Sodium Chloride, 5gSold as 2 x 500g.
Catalog No. 50-254-409 Supplier Bioworld Supplier No. 306201051
Used for the cultivation of recombinant strains of Escherichia coli. Composition per liter:Bacto Tryptone, 8gBacto Yeast Extract, 5g Sodium Chloride, 5gSold as 2 x 500g.