Learn More
Cayman Chemical YTHDC2 PolycLNal ANTIBDY 500ug
SDP
Catalog No. 502941806
Encompass_Preferred
Immunogen: Full-length recombinant human YTHDC2 protein • Host: Rabbit • Species Reactivity: (+) Human • Applications: IF, IHC, and WB • MW = 160 kDa
Catalog No. 50-294-1806 Supplier Cayman Chemical Supplier No. 28224500 UG
Immunogen: Full-length recombinant human YTHDC2 protein • Host: Rabbit • Species Reactivity: (+) Human • Applications: IF, IHC, and WB • MW = 160 kDa